C12H316CEC0D Chemcia Scientific, LLC

3-Trifluoromethyl-4,5,6,7-tetrahydro-1H-indazole

  • MDL number MFCD01312181
  • Cas Number : 35179-55-6
  • Molecular Weight : 190.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-(trifluoromethyl)-4,5,6,7-tetrahydro-1H-indazole
Inchi Code 1S/C8H9F3N2/c9-8(10,11)7-5-3-1-2-4-6(5)12-13-7/h1-4H2,(H,12,13)
Inchi Key KTULFINKQNWXNY-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3763

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3763
Availability details for
Quantity
  • Availability
  • Add to Cart