C12H316CEC0F Chemcia Scientific, LLC

(5-Methoxy-1H-benzoimidazol-2-yl)-methanol

  • MDL number MFCD01073114
  • Cas Number : 20033-99-2
  • Molecular Weight : 178.19225

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3765

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3765
Availability details for
Quantity
  • Availability
  • Add to Cart