C12H316CEC12 Chemcia Scientific, LLC

(4,5,6,7-Tetrahydro-2H-indazol-5-yl)-carbamic acid tert-butyl ester

  • MDL number MFCD28044185
  • Cas Number : 955406-11-8
  • Molecular Weight : 237.30413

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3768

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3768
Availability details for
Quantity
  • Availability
  • Add to Cart