C12H316CEC14 Chemcia Scientific, LLC

5-Bromo-2-fluoro-4-methoxy-benzoic acid

  • MDL number MFCD20486270
  • Cas Number : 949014-42-0
  • Molecular Weight : 249.03762

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3770

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3770
Availability details for
Quantity
  • Availability
  • Add to Cart