C12H316CEC1C Chemcia Scientific, LLC

4-Amino-1-benzyl-1,2,5,6-tetrahydro-pyridine-3-carboxylic acid ethyl ester

  • MDL number MFCD11867863
  • Cas Number : 70336-83-3
  • Molecular Weight : 260.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 4-amino-1-benzyl-1,2,5,6-tetrahydro-3-pyridinecarboxylate
Inchi Code 1S/C15H20N2O2/c1-2-19-15(18)13-11-17(9-8-14(13)16)10-12-6-4-3-5-7-12/h3-7H,2,8-11,16H2,1H3
Inchi Key ULARACKZXVBXJF-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3778

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3778
Availability details for
Quantity
  • Availability
  • Add to Cart