C12H316CEC1F Chemcia Scientific, LLC

(2-Bromo-5-fluoro-phenyl)-methyl-amine

  • MDL number MFCD09745014
  • Cas Number : 1200114-00-6
  • Molecular Weight : 204.04294

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3781

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3781
Availability details for
Quantity
  • Availability
  • Add to Cart