C12H316CEC20 Chemcia Scientific, LLC

2-(2,2-Dimethoxy-ethyl)-phenylamine

  • MDL number MFCD09833913
  • Cas Number : 150760-45-5
  • Molecular Weight : 181.23655

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Brown Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3782

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3782
Availability details for
Quantity
  • Availability
  • Add to Cart