C12H316CEC22 Chemcia Scientific, LLC

2-Fluoro-5-nitro-benzamide

  • MDL number MFCD12168113
  • Cas Number : 136146-82-2
  • Molecular Weight : 184.1279

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3784

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3784
Availability details for
Quantity
  • Availability
  • Add to Cart