C12H316CEC2A Chemcia Scientific, LLC

2,5-Dibromo-4-nitro-pyridine 1-oxide

  • MDL number MFCD00233996
  • Cas Number : 221241-25-4
  • Molecular Weight : 297.89129

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3792

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3792
Availability details for
Quantity
  • Availability
  • Add to Cart