C12H316CEC2C Chemcia Scientific, LLC

(3-Bromo-5-nitro-pyridin-2-yl)-hydrazine

  • MDL number MFCD01117228
  • Cas Number : 15862-38-1
  • Molecular Weight : 233.02

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 3-bromo-2-hydrazino-5-nitropyridine
Inchi Code 1S/C5H5BrN4O2/c6-4-1-3(10(11)12)2-8-5(4)9-7/h1-2H,7H2,(H,8,9)
Inchi Key FGRGIVXZHDLCBP-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3794

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3794
Availability details for
Quantity
  • Availability
  • Add to Cart