C12H316CEC2F Chemcia Scientific, LLC

1-(2-Amino-benzothiazol-6-yl)-ethanone

  • MDL number MFCD00548461
  • Cas Number : 21222-61-7
  • Molecular Weight : 192.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 1-(2-amino-1,3-benzothiazol-6-yl)ethanone
Inchi Code 1S/C9H8N2OS/c1-5(12)6-2-3-7-8(4-6)13-9(10)11-7/h2-4H,1H3,(H2,10,11)
Inchi Key OKAQYTQTHTXQQH-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3797

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3797
Availability details for
Quantity
  • Availability
  • Add to Cart