C12H316CEC30 Chemcia Scientific, LLC

Hexahydro-oxazolo[3,4-a]pyridin-3-one

  • MDL number MFCD13179087
  • Cas Number : 42329-17-9
  • Molecular Weight : 141.17122

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3798

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3798
Availability details for
Quantity
  • Availability
  • Add to Cart