C12H316CEC31 Chemcia Scientific, LLC

2,4,6,7-Tetrahydro-pyrano[4,3-c]pyrazole-3-carboxylic acid

  • MDL number MFCD1520226
  • Cas Number : 933694-89-4
  • Molecular Weight : 168.15341

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3799

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3799
Availability details for
Quantity
  • Availability
  • Add to Cart