C12H316CEC32 Chemcia Scientific, LLC

(4,5,6,7-Tetrahydro-1H-indazol-5-yl)-carbamic acid tert-butyl ester

  • MDL number MFCD12408337
  • Cas Number : 1158767-01-1
  • Molecular Weight : 237.30413

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3800

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3800
Availability details for
Quantity
  • Availability
  • Add to Cart