C12H316CEC34 Chemcia Scientific, LLC

(6-Fluoro-pyridin-3-yl)-dimethyl-amine

  • MDL number MFCD28154575
  • Cas Number : 1823624-32-3
  • Molecular Weight : 140.16158

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3805

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3805
Availability details for
Quantity
  • Availability
  • Add to Cart