C12H316CEC36 Chemcia Scientific, LLC

5-Fluoro-pyridine-2-carboxylic acid ethyl ester

  • MDL number MFCD11616895
  • Cas Number : 148541-70-2
  • Molecular Weight : 169.15686

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3812

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3812
Availability details for
Quantity
  • Availability
  • Add to Cart