C12H316CEC3A Chemcia Scientific, LLC

(6-Fluoro-pyridin-3-yl)-methyl-amine

  • MDL number MFCD28154576
  • Cas Number : 1610667-14-5
  • Molecular Weight : 126.13449

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light yellow solid at 0 degree C
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3816

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3816
Availability details for
Quantity
  • Availability
  • Add to Cart