C12H316CEC3C Chemcia Scientific, LLC

2-Amino-benzothiazole-6-carbaldehyde

  • MDL number MFCD11976759
  • Cas Number : 106429-08-7
  • Molecular Weight : 178.21382

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3821

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3821
Availability details for
Quantity
  • Availability
  • Add to Cart