C12H316CEC3F Chemcia Scientific, LLC

4-Bromo-2H-indazol-3-ylamine

  • MDL number MFCD27989048
  • Cas Number : 914311-50-5
  • Molecular Weight : 212.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 4-bromo-2H-indazol-3-amine
Inchi Code 1S/C7H6BrN3/c8-4-2-1-3-5-6(4)7(9)11-10-5/h1-3H,(H3,9,10,11)
Inchi Key IXXXOIXGQFPLIH-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Red Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3832

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3832
Availability details for
Quantity
  • Availability
  • Add to Cart