C12H316CEC41 Chemcia Scientific, LLC

7-Fluoro-2H-indazol-3-ylamine

  • MDL number MFCD28154578
  • Cas Number : 404827-60-7
  • Molecular Weight : 151.14437

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3834

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3834
Availability details for
Quantity
  • Availability
  • Add to Cart