C12H316CEC43 Chemcia Scientific, LLC

2-Fluoro-5-hydroxy-benzamide

  • MDL number MFCD16999275
  • Cas Number : 951122-32-0
  • Molecular Weight : 155.12977

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3840

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3840
Availability details for
Quantity
  • Availability
  • Add to Cart