C12H316CEC47 Chemcia Scientific, LLC

7-Methoxy-imidazo[1,2-a]pyridine

  • MDL number MFCD13185227
  • Cas Number : 342613-71-2
  • Molecular Weight : 148.16576

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Brown Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3850

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3850
Availability details for
Quantity
  • Availability
  • Add to Cart