C12H316CEC4A Chemcia Scientific, LLC

8-Bromo-pyrido[1,2-a]pyrimidin-4-one

  • MDL number MFCD25956423
  • Cas Number : 1198413-10-3
  • Molecular Weight : 225.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 8-bromo-4H-pyrido[1,2-a]pyrimidin-4-one
Inchi Code 1S/C8H5BrN2O/c9-6-2-4-11-7(5-6)10-3-1-8(11)12/h1-5H
Inchi Key CVYFPYSSTYXLMX-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3857

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3857
Availability details for
Quantity
  • Availability
  • Add to Cart