C12H316CEC4B Chemcia Scientific, LLC

8-Chloro-[1,2,4]triazolo[1,5-a]pyridine

  • MDL number MFCD23708544
  • Cas Number : 1427368-62-4
  • Molecular Weight : 153.57188

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3860

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3860
Availability details for
Quantity
  • Availability
  • Add to Cart