C12H316CEC5A Chemcia Scientific, LLC

(2-Fluoro-5-methoxy-phenyl)-acetonitrile

  • MDL number MFCD09038466
  • Cas Number : 672931-28-1
  • Molecular Weight : 165.16861

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3880

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3880
Availability details for
Quantity
  • Availability
  • Add to Cart