C12H316CEC5E Chemcia Scientific, LLC

5-Chloro-6-fluoro-pyridin-2-ylamine

  • MDL number MFCD18253522
  • Cas Number : 1378595-22-2
  • Molecular Weight : 146.55243

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Pink Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3884

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3884
Availability details for
Quantity
  • Availability
  • Add to Cart