C12H316CEC5F Chemcia Scientific, LLC

4-Methyl-3H-benzothiazol-2-one

  • MDL number MFCD17178463
  • Cas Number : 73443-84-2
  • Molecular Weight : 165.21509

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3885

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3885
Availability details for
Quantity
  • Availability
  • Add to Cart