C12H316CEC65 Chemcia Scientific, LLC

2-Cyclopropyl-5,6,7,8-tetrahydro-pyrido[4,3-d]pyrimidine

  • MDL number MFCD06656228
  • Cas Number : 880361-75-1
  • Molecular Weight : 175.23521

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3891

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3891
Availability details for
Quantity
  • Availability
  • Add to Cart