C12H316CEC6A Chemcia Scientific, LLC

6-Bromo-imidazo[1,2-a]pyridine-3-carbonitrile

  • MDL number MFCD11858502
  • Cas Number : 474708-98-0
  • Molecular Weight : 222.04

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-bromoimidazo[1,2-a]pyridine-3-carbonitrile
Inchi Code 1S/C8H4BrN3/c9-6-1-2-8-11-4-7(3-10)12(8)5-6/h1-2,4-5H
Inchi Key PJSBOEVNEUCHIG-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3896

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3896
Availability details for
Quantity
  • Availability
  • Add to Cart