C12H316CEC6E Chemcia Scientific, LLC

5-Methoxy-3-methyl-pyridine-2-carbonitrile

  • MDL number MFCD18255402
  • Cas Number : 1256792-12-7
  • Molecular Weight : 148.16576

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Beige Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3900

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3900
Availability details for
Quantity
  • Availability
  • Add to Cart