C12H316CEC87 Chemcia Scientific, LLC

(5-Bromo-pyrazin-2-yl)-methyl-amine

  • MDL number MFCD09881407
  • Cas Number : 446286-92-6
  • Molecular Weight : 188.02767

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3934

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3934
Availability details for
Quantity
  • Availability
  • Add to Cart