C12H316CEC8A Chemcia Scientific, LLC

Dimethyl-(4-methyl-pyridin-2-yl)-amine

  • MDL number MFCD17015432
  • Cas Number : 20173-72-2
  • Molecular Weight : 136.19824

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3937

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3937
Availability details for
Quantity
  • Availability
  • Add to Cart