C12H316CEC8D Chemcia Scientific, LLC

3-Iodo-4-methyl-pyridin-2-ylamine

  • MDL number MFCD16606186
  • Cas Number : 1227509-37-6
  • Molecular Weight : 234.04049

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form Off-White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3940

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3940
Availability details for
Quantity
  • Availability
  • Add to Cart