C12H316CEC8F Chemcia Scientific, LLC

2-(2-Methoxy-phenyl)-propan-2-ol

  • MDL number MFCD12068167
  • Cas Number : 21022-73-1
  • Molecular Weight : 166.22188

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3942

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3942
Availability details for
Quantity
  • Availability
  • Add to Cart