C12H316CEC92 Chemcia Scientific, LLC

3-Bromo-5-fluoro-imidazo[1,2-a]pyridine

  • MDL number MFCD22372500
  • Cas Number : 1388711-44-1
  • Molecular Weight : 215.02573

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Tan Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3945

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3945
Availability details for
Quantity
  • Availability
  • Add to Cart