C12H316CEC93 Chemcia Scientific, LLC

Dimethyl-(3-methyl-pyridin-2-yl)-amine

  • MDL number MFCD27979104
  • Cas Number : 61713-46-0
  • Molecular Weight : 136.19824

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3946

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3946
Availability details for
Quantity
  • Availability
  • Add to Cart