C12H316CEC9F Chemcia Scientific, LLC

(2,5-Difluoro-phenyl)-methyl-amine

  • MDL number MFCD11151819
  • Cas Number : 138563-58-3
  • Molecular Weight : 143.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2,5-difluoro-N-methylaniline
Inchi Code 1S/C7H7F2N/c1-10-7-4-5(8)2-3-6(7)9/h2-4,10H,1H3
Inchi Key SAGAEZGLJJTKRE-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=3959

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3959
Availability details for
Quantity
  • Availability
  • Add to Cart