C12H316CECA0 Chemcia Scientific, LLC

(2,5-Difluoro-phenyl)-carbamic acid tert-butyl ester

  • MDL number MFCD21170328
  • Cas Number : 129589-71-5
  • Molecular Weight : 229.22856

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Orange Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=3960

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=3960
Availability details for
Quantity
  • Availability
  • Add to Cart