C12H316CECAE Chemcia Scientific, LLC

(2-Chloro-benzothiazol-6-yl)-acetic acid ethyl ester

  • MDL number MFCD28154599
  • Cas Number : 1823505-06-1
  • Molecular Weight : 255.72

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name ethyl 2-(2-chlorobenzo[d]thiazol-6-yl)acetate
Inchi Code 1S/C11H10ClNO2S/c1-2-15-10(14)6-7-3-4-8-9(5-7)16-11(12)13-8/h3-5H,2,6H2,1H3
Inchi Key LDSBFLRPTGHFAC-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=4061

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4061
Availability details for
Quantity
  • Availability
  • Add to Cart