C12H316CECAF Chemcia Scientific, LLC

6-Chloro-imidazo[1,2-a]pyridine-3-carbonitrile

  • MDL number MFCD11858503
  • Cas Number : 123531-24-8
  • Molecular Weight : 177.59

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 6-chloroimidazo[1,2-a]pyridine-3-carbonitrile
Inchi Code 1S/C8H4ClN3/c9-6-1-2-8-11-4-7(3-10)12(8)5-6/h1-2,4-5H
Inchi Key PZIUWGQZDDJQMW-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form Brown Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=4062

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4062
Availability details for
Quantity
  • Availability
  • Add to Cart