C12H316CECB1 Chemcia Scientific, LLC

2-Bromo-8-chloro-[1,2,4]triazolo[1,5-a]pyridine

  • MDL number MFCD28154600
  • Cas Number : 1784176-25-5
  • Molecular Weight : 232.47

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 2-bromo-8-chloro-[1,2,4]triazolo[1,5-a]pyridine
Inchi Code 1S/C6H3BrClN3/c7-6-9-5-4(8)2-1-3-11(5)10-6/h1-3H
Inchi Key CQJRTZCZDYDQPO-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=4064

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4064
Availability details for
Quantity
  • Availability
  • Add to Cart