C12H316CECCF Chemcia Scientific, LLC

5-Chlorosulfonyl-2,4-difluoro-benzoic acid

  • MDL number MFCD09050371
  • Cas Number : 53916-70-4
  • Molecular Weight : 256.61336

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4071

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4071
Availability details for
Quantity
  • Availability
  • Add to Cart