C12H316CECD0 Chemcia Scientific, LLC

5-Fluoro-7-nitro-1H-indole-2,3-dione

  • MDL number MFCD09742036
  • Cas Number : 954571-39-2
  • Molecular Weight : 210.12

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 5-fluoro-7-nitro-1H-indole-2,3-dione
Inchi Code 1S/C8H3FN2O4/c9-3-1-4-6(5(2-3)11(14)15)10-8(13)7(4)12/h1-2H,(H,10,12,13)
Inchi Key WGBLCTOGPSHGGJ-UHFFFAOYSA-N

Description

Purity 95%
Storage temp. 0~5
Physical Form Yellow Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=4072

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4072
Availability details for
Quantity
  • Availability
  • Add to Cart