C12H316CECD3 Chemcia Scientific, LLC

(4-Bromo-phenyl)-cyclopentyl-acetic acid

  • MDL number MFCD11590509
  • Cas Number : 1082453-57-3
  • Molecular Weight : 283.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name (4-bromophenyl)(cyclopentyl)acetic acid
Inchi Code 1S/C13H15BrO2/c14-11-7-5-10(6-8-11)12(13(15)16)9-3-1-2-4-9/h5-9,12H,1-4H2,(H,15,16)
Inchi Key QXBDOMRNBZRJKA-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=4075

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4075
Availability details for
Quantity
  • Availability
  • Add to Cart