C12H316CECD4 Chemcia Scientific, LLC

7-Chloro-1H-indazol-3-ylamine

  • MDL number MFCD07385618
  • Cas Number : 88805-67-8
  • Molecular Weight : 167.59897

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4076

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4076
Availability details for
Quantity
  • Availability
  • Add to Cart