C12H316CECD5 Chemcia Scientific, LLC

7-Chloro-2H-indazol-3-ylamine

  • MDL number MFCD28154601
  • Cas Number : 88805-67-8
  • Molecular Weight : 167.6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Properties

Iupac Name 7-chloro-2H-indazol-3-ylamine
Inchi Code 1S/C7H6ClN3/c8-5-3-1-2-4-6(5)10-11-7(4)9/h1-3H,(H3,9,10,11)
Inchi Key HNISSRAYDSNHNN-UHFFFAOYSA-N

Description

Purity 97%
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin US
Product Link http://www.chemcia.com/product_fl02.asp?id=4077

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4077
Availability details for
Quantity
  • Availability
  • Add to Cart