C12H316CECE1 Chemcia Scientific, LLC

2,4-Difluoro-benzoic acid tert-butyl ester

  • MDL number MFCD19388670
  • Cas Number : 500353-15-1
  • Molecular Weight : 214.21389

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4080

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4080
Availability details for
Quantity
  • Availability
  • Add to Cart