C12H316CECE7 Chemcia Scientific, LLC

2-Amino-4-bromo-5-iodo-benzoic acid

  • MDL number MFCD28154603
  • Cas Number : 1823494-61-6
  • Molecular Weight : 341.9318

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Light Yellow Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4159

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4159
Availability details for
Quantity
  • Availability
  • Add to Cart