C12H316CECF6 Chemcia Scientific, LLC

Methyl-(4-methyl-pyridin-2-yl)-amine

  • MDL number MFCD00956049
  • Cas Number : 45699-12-5
  • Molecular Weight : 122.17115

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 0~5
Physical Form Yellow Liquid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4160

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4160
Availability details for
Quantity
  • Availability
  • Add to Cart