C12H316CECFA Chemcia Scientific, LLC

7-Chloro-imidazo[1,2-a]pyridine-2-carboxylic acid ethyl ester hydrobromide

  • MDL number MFCD28155286
  • Cas Number : 1383080-55-4
  • Molecular Weight : 305.5604

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 97
Salt Data HBr
Storage temp. 0~5
Physical Form White Solid
Shipping Temperature Ambient
Country of Origin China
Product Link http://www.chemcia.com/product_fl02.asp?id=4164

Safety Information

MSDS http://www.chemcia.com/product_fl02.asp?id=4164
Availability details for
Quantity
  • Availability
  • Add to Cart